APO455827665 Apollo Scientific Ltd

1-[2-Chloro-4-(4-chlorophenoxy)phenyl]ethan-1-one

  • MDL number MFCD00140226
  • Cas Number : 119851-28-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 54-58
Synonym 2'-Chloro-4'-(4-chlorophenoxy)acetophenone, 4-Acetyl-3,4'-dichlorodiphenyl ether
Purity 98
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/1-2-chloro-4-4-chlorophenoxyphenylethan-1-one

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/1-2-chloro-4-4-chlorophenoxyphenylethan-1-one
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart