APO455827676 Apollo Scientific Ltd

5-Methyl-3-phenyl-1H-pyrazole

  • MDL number MFCD00524752
  • Cas Number : 3347-62-4
  • Molecular Weight : 158.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 124-127
Iupac Name 5-methyl-3-phenyl-1H-pyrazole
Inchi Code 1S/C10H10N2/c1-8-7-10(12-11-8)9-5-3-2-4-6-9/h2-7H,1H3,(H,11,12)
Inchi Key QHRSESMSOJZMCO-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 124-127
Product Link https://store.apolloscientific.co.uk/product/5-methyl-3-phenyl-1h-pyrazole

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/5-methyl-3-phenyl-1h-pyrazole
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart