APO455827677 Apollo Scientific Ltd

2-(2-Oxo-1,3-oxazolidin-3-yl)ethyl toluene-4-sulphonate

  • MDL number MFCD08276953
  • Cas Number : 159974-55-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 68-69
Synonym 2-(2-Oxo-1,3-oxazolidin-3-yl)ethyl 4-methylbenzenesulphonate, 2-(2-Oxo-1,3-oxazolidin-3-yl)ethyl tosylate
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=39672

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=39672
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart