APO455827678 Apollo Scientific Ltd

1-Methyl-5-[(methylamino)methyl]-1H-pyrazole

  • MDL number MFCD03419812
  • Cas Number : 930111-04-9
  • Molecular Weight : 125.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N-methyl(1-methyl-1H-pyrazol-5-yl)methanamine
Inchi Code 1S/C6H11N3/c1-7-5-6-3-4-8-9(6)2/h3-4,7H,5H2,1-2H3
Inchi Key CHGIPIYKGPNYKT-UHFFFAOYSA-N

Description

Synonym N-Methyl-1-(1-methyl-1H-pyrazol-5-yl)methylamine
Purity 97%
Flash Point 102/15mm
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/1-methyl-5-methylaminomethyl-1h-pyrazole

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/1-methyl-5-methylaminomethyl-1h-pyrazole
pictograms GHS05;GHS07
hstatements H302;H314
Availability details for
Quantity
  • Availability
  • Add to Cart