APO455827679 Apollo Scientific Ltd

3-Bromo-4-[(methylamino)methyl]thiophene hydrochloride

  • MDL number MFCD09817483
  • Cas Number : 944450-82-2
  • Molecular Weight : 242.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 164.5-166
Iupac Name (4-bromo-3-thienyl)-N-methylmethanamine hydrochloride
Inchi Code 1S/C6H8BrNS.ClH/c1-8-2-5-3-9-4-6(5)7;/h3-4,8H,2H2,1H3;1H
Inchi Key GORGMQYTNXIOBQ-UHFFFAOYSA-N

Description

Synonym N-Methyl-[(4-bromothien-3-yl)methyl]amine hydrochloride
Storage temp. Store at 2-8
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 164.5-166
Product Link https://store.apolloscientific.co.uk/product/3-bromo-4-methylaminomethylthiophene-hydrochloride

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/3-bromo-4-methylaminomethylthiophene-hydrochloride
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart