APO455827680 Apollo Scientific Ltd

2-Phenylpyrimidine-5-carbonyl chloride

  • MDL number MFCD09817549
  • Cas Number : 122774-00-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 174-176
Synonym 5-(Chlorocarbonyl)-2-phenylpyrimidine
Purity 97
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=39771

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=39771
pictograms GHS05
hstatements H314;EUH029;EUH014
Availability details for
Quantity
  • Availability
  • Add to Cart