APO455827689 Apollo Scientific Ltd

5-(4-Chlorophenyl)thiophene-2-carboxaldehyde

  • MDL number MFCD01860767
  • Cas Number : 38401-71-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 85-86
Synonym 4-(2-Formylthiophene)chlorobenzene
Storage temp. Ambient temperature
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=39821

Safety Information

MSDS http://www.apolloscientific.co.uk/display_item.php?id=39821
Availability details for
Quantity
  • Availability
  • Add to Cart