APO455827694 Apollo Scientific Ltd

Methyl 2-amino-3-methylbenzoate

  • MDL number MFCD06200918
  • Cas Number : 22223-49-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-(Methoxycarbonyl)-6-methylaniline, 2-Amino-3-(methoxycarbonyl)toluene, Methyl 3-methylanthranilate
Purity 98
Storage temp. Store at 2-8
Physical Form Liquid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=39858

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=39858
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart