APO455827696 Apollo Scientific Ltd

Methyl 1-(4,6-dimethylpyrimidin-2-yl)-1H-1,2,3-triazole-4-carboxylate

  • MDL number MFCD00838109
  • Cas Number : 23947-13-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 184-186
Synonym 1-(4,6-Dimethylpyrimidin-2-yl)-4-(methoxycarbonyl)-1H-1,2,3-triazole, 4,6-Dimethyl-2-[(4-methoxycarbonyl)-1H-1,2,3-triazol-1-yl]pyrimidine
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=39860

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=39860
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart