APO455827700 Apollo Scientific Ltd

[3-(4-Chlorophenyl)-1,2,4-oxadiazol-5-yl]methanol

  • MDL number MFCD06200919
  • Cas Number : 5372-40-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 96-99
Synonym 3-(4-Chlorophenyl)-5-(hydroxymethyl)-1,2,4-oxadiazole
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=39950

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=39950
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart