APO455827701 Apollo Scientific Ltd

4,5-Dibromo-1H-pyrrole-2-carboxylic acid

  • MDL number MFCD01348229
  • Cas Number : 34649-21-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point >300
Synonym 5-Carboxy-2,3-dibromo-1H-pyrrole
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=39952

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=39952
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart