APO455827708 Apollo Scientific Ltd

4-Propanoyl-1H-pyrrole-2-carboxylic acid

  • MDL number MFCD06200986
  • Cas Number : 111468-94-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 216-220
Synonym 2-Carboxy-4-(ethylcarbonyl)-1H-pyrrole, 4-Propionyl-1H-pyrrole-2-carboxylic acid
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=40019

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=40019
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart