APO455827710 Apollo Scientific Ltd

5-Bromo-2-(hydroxymethyl)-1-methyl-1H-imidazole

  • MDL number MFCD09864833
  • Cas Number : 861362-06-3
  • Molecular Weight : 191.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 143-145
Synonym (5-Bromo-1-methyl-1H-imidazol-2-yl)methanol
Purity 97
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-bromo-2-hydroxymethyl-1-methyl-1h-imidazole

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-bromo-2-hydroxymethyl-1-methyl-1h-imidazole
pictograms GHS07
hstatements H302+H312+H332
Availability details for
Quantity
  • Availability
  • Add to Cart