APO455827713 Apollo Scientific Ltd

4',5'-Dihydro-1H,3'H-2,2'-bipyrrole

  • MDL number MFCD00030437
  • Cas Number : 10087-65-7
  • Molecular Weight : 134.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 162-166
Synonym 2-(3,4-Dihydro-2H-pyrrol-5-yl)-1H-pyrrole, 3,4-Dihydro-5-(1H-pyrrol-2-yl)-2H-pyrrole
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/45-dihydro-1h3h-22-bipyrrole

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/45-dihydro-1h3h-22-bipyrrole
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart