APO455827715 Apollo Scientific Ltd

Methyl trans-4-bromocinnamate

  • MDL number MFCD00157162
  • Cas Number : 71205-17-9
  • Molecular Weight : 241.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 84-86
Synonym Methyl (E)-3-(4-bromophenyl)acrylate, Methyl (2E)-3-(4-bromophenyl)prop-2-enoate
Purity 98
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/methyl-trans-4-bromocinnamate

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-trans-4-bromocinnamate
pictograms GHS07
hstatements H302+H312+H332
Availability details for
Quantity
  • Availability
  • Add to Cart