APO455827718 Apollo Scientific Ltd

4-Amino-5-bromo-2-chloropyrimidine

  • MDL number MFCD07782040
  • Cas Number : 205672-25-9
  • Molecular Weight : 208.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 189-191
Iupac Name 5-bromo-2-chloro-4-pyrimidinylamine
Inchi Code 1S/C4H3BrClN3/c5-2-1-8-4(6)9-3(2)7/h1H,(H2,7,8,9)
Inchi Key QOWALNIZDHZTSM-UHFFFAOYSA-N

Description

Synonym 5-Bromo-2-chloropyrimidin-4-amine, 4-Amino-5-bromo-2-chloro-1,3-diazine
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 189-191
Product Link https://store.apolloscientific.co.uk/product/4-amino-5-bromo-2-chloropyrimidine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-amino-5-bromo-2-chloropyrimidine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart