APO455827722 Apollo Scientific Ltd

2-Chloro-4-(methylthio)pyrimidine

  • MDL number MFCD09265490
  • Cas Number : 49844-93-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 59-64
Synonym 2-Chloro-4-(methylsulphanyl)pyrimidine, 2-Chloro-4-(methylsulphonyl)-1,3-diazine
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/2-chloro-4-methylthiopyrimidine

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-chloro-4-methylthiopyrimidine
pictograms GHS05;GHS07
hstatements H314;H335
Availability details for
Quantity
  • Availability
  • Add to Cart