APO455827726 Apollo Scientific Ltd

2-(Methylthio)pyrimidine-5-boronic acid

  • MDL number MFCD07375142
  • Cas Number : 348098-29-3
  • Molecular Weight : 170

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-(Methylsulfanyl)pyrimidine-5-boronic acid, 2-(Methylsulphanyl)pyrimidine-5-boronic acid
Purity 97
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-methylthiopyrimidine-5-boronic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-methylthiopyrimidine-5-boronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart