APO455827734 Apollo Scientific Ltd

5-(Tributylstannyl)-1,3-thiazole

  • MDL number MFCD01319074
  • Cas Number : 157025-33-7
  • Molecular Weight : 374.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-(tributylstannyl)-1,3-thiazole
Inchi Code 1S/3C4H9.C3H2NS.Sn/c3*1-3-4-2;1-2-5-3-4-1;/h3*1,3-4H2,2H3;1,3H;
Inchi Key GOWNSHUNTJWVOM-UHFFFAOYSA-N

Description

Synonym (1,3-Thiazol-5-yl)tributylstannane
Storage temp. Store at 2-8
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-tributylstannyl-13-thiazole

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-tributylstannyl-13-thiazole
pictograms GHS06;GHS08;GHS09
hstatements H301;H312;H319;H410
Availability details for
Quantity
  • Availability
  • Add to Cart