APO455827735 Apollo Scientific Ltd

2-Acetamido-5-bromo-1,3-thiazole

  • MDL number MFCD00017336
  • Cas Number : 7336-54-1
  • Molecular Weight : 221.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 198-201
Iupac Name N-(5-bromo-1,3-thiazol-2-yl)acetamide
Inchi Code 1S/C5H5BrN2OS/c1-3(9)8-5-7-2-4(6)10-5/h2H,1H3,(H,7,8,9)
Inchi Key LHWHLINDRWCHSN-UHFFFAOYSA-N

Description

Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 198-201
Product Link https://store.apolloscientific.co.uk/product/2-acetamido-5-bromo-13-thiazole

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/2-acetamido-5-bromo-13-thiazole
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart