APO455827738 Apollo Scientific Ltd

4-Bromo-2-ethoxy-1,3-thiazole

  • MDL number MFCD07787375
  • Cas Number : 240816-34-6
  • Molecular Weight : 208.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-Bromo-1,3-thiazol-2-yl methyl ether
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-bromo-2-ethoxy-13-thiazole

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/4-bromo-2-ethoxy-13-thiazole
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart