APO455827752 Apollo Scientific Ltd

5-(Hydroxymethyl)-4-methyl-1H-imidazole hydrochloride

  • MDL number MFCD00012696
  • Cas Number : 38585-62-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 233
Synonym (4-Methyl-1H-imidazol-5-yl)methanol hydrochloride
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/5-hydroxymethyl-4-methyl-1h-imidazole-hydrochloride

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/5-hydroxymethyl-4-methyl-1h-imidazole-hydrochloride
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart