APO455827753 Apollo Scientific Ltd

3-Chloro-2-methylphenol

  • MDL number MFCD00079738
  • Cas Number : 3260-87-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 86
Synonym 2-Chloro-6-hydroxytoluene, 3-Chloro-o-cresol
Purity 98
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Boiling point 225
Product Link http://www.apolloscientific.co.uk/display_item.php?id=29172

Safety Information

signalword Danger
pcodes P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=29172
pictograms GHS07;GHS05
hstatements H302;H312;H318
Availability details for
Quantity
  • Availability
  • Add to Cart