APO455827759 Apollo Scientific Ltd

1-[Tris(isopropyl)silyl]-1H-pyrrole-3-boronic acid

  • MDL number MFCD01114667
  • Cas Number : 138900-55-7
  • Molecular Weight : 267.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(triisopropylsilyl)-1H-pyrrol-3-ylboronic acid
Inchi Code 1S/C13H26BNO2Si/c1-10(2)18(11(3)4,12(5)6)15-8-7-13(9-15)14(16)17/h7-12,16-17H,1-6H3
Inchi Key HUBVAOMVEMGRFA-UHFFFAOYSA-N

Description

Synonym 3-Borono-1-[tris(isopropyl)silyl]-1H-pyrrole, (3-Borono-1H-pyrrol-1-yl)[tris(prop-2-yl)]silane, (3-Borono-1H-pyrol-1-yl)[tris(isopropyl)]silane
Purity 98%
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/1-trisisopropylsilyl-1h-pyrrole-3-boronic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/1-trisisopropylsilyl-1h-pyrrole-3-boronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart