APO455827778 Apollo Scientific Ltd

3-Methylpiperazine, N1-BOC protected

  • MDL number MFCD03001706
  • Cas Number : 120737-59-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym tert-Butyl 3-methylpiperazine-1-carboxylate, 1-(tert-Butoxycarbonyl)-3-methylpiperazine
Purity 97
Storage temp. Ambient temperature
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=42766

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=42766
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart