APO455827783 Apollo Scientific Ltd

(2S)-2-Ethylpiperazine, N1-BOC protected

  • MDL number MFCD07772100
  • Cas Number : 325145-35-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym tert-Butyl (2S)-2-ethylpiperazine-1-carboxylate, (2S)-1-(tert-Butoxycarbonyl)-2-ethylpiperazine
Storage temp. Store at 2-8
Physical Form Liquid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=42784

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=42784
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart