APO455827786 Apollo Scientific Ltd

1-Cyclopropylpiperazine dihydrochloride

  • MDL number MFCD00040810
  • Cas Number : 139256-79-4
  • Molecular Weight : 199.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-cyclopropylpiperazine dihydrochloride
Inchi Code 1S/C7H14N2.2ClH/c1-2-7(1)9-5-3-8-4-6-9;;/h7-8H,1-6H2;2*1H
Inchi Key JDJNSFSTSJWJFA-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/1-cyclopropylpiperazine-dihydrochloride

Safety Information

signalword Danger
pcodes P260;P280;P310
MSDS https://store.apolloscientific.co.uk/product/1-cyclopropylpiperazine-dihydrochloride
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart