APO455827787 Apollo Scientific Ltd

1-Cyclopropylpiperazine

  • MDL number MFCD06254805
  • Cas Number : 20327-23-5
  • Molecular Weight : 126.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-cyclopropylpiperazine
density 0.94
Inchi Code 1S/C7H14N2/c1-2-7(1)9-5-3-8-4-6-9/h7-8H,1-6H2
Inchi Key HNZJIWIXRPBFAN-UHFFFAOYSA-N

Description

Density 0.94
Flash Point 43/7mm
Storage temp. Store at 2-8
Physical Form Solid-Low Melt
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/1-cyclopropylpiperazine

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/1-cyclopropylpiperazine
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart