APO455827795 Apollo Scientific Ltd

(3S)-3-(Hydroxymethyl)piperidine, N-BOC protected

  • MDL number MFCD02683203
  • Cas Number : 140695-84-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym tert-Butyl (3S)-3-(hydroxymethyl)piperidine-1-carboxylate, (3S)-1-(tert-Butoxycarbonyl)-3-(hydroxymethyl)piperidine
Storage temp. Ambient temperature
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=43214

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=43214
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart