APO455827801 Apollo Scientific Ltd

(3S)-3-Hydroxypyrrolidine hydrochloride

  • MDL number MFCD00272298
  • Cas Number : 122536-94-1
  • Molecular Weight : 123.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 106-107
Iupac Name (3S)-3-pyrrolidinol hydrochloride
Inchi Code 1S/C4H9NO.ClH/c6-4-1-2-5-3-4;/h4-6H,1-3H2;1H/t4-;/m0./s1
Inchi Key QPMSJEFZULFYTB-WCCKRBBISA-N

Description

Synonym (3S)-Pyrrolidin-3-ol hydrochloride
Flash Point 108-110
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 106-107
Product Link https://store.apolloscientific.co.uk/product/3s-3-hydroxypyrrolidine-hydrochloride

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3s-3-hydroxypyrrolidine-hydrochloride
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart