APO455827806 Apollo Scientific Ltd

Azetidine-3-carboxaldehyde, N-BOC protected

  • MDL number MFCD06656142
  • Cas Number : 177947-96-5
  • Molecular Weight : 185.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl 3-formyl-1-azetidinecarboxylate
Inchi Code 1S/C9H15NO3/c1-9(2,3)13-8(12)10-4-7(5-10)6-11/h6-7H,4-5H2,1-3H3
Inchi Key JVQOZRRUGOADSU-UHFFFAOYSA-N

Description

Synonym tert-Butyl 3-formylazetidine-1-carboxylate, N-(tert-Butoxycarbonyl)azetane-3-carboxaldehyde
Flash Point 77-82/2mm
Storage temp. Store at -20
Physical Form Liquid
Shipping Temperature Shipped with Ice Packs
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/azetidine-3-carboxaldehyde-n-boc-protected

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/azetidine-3-carboxaldehyde-n-boc-protected
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart