APO455827815 Apollo Scientific Ltd

7-Bromobenzo[b]furan

  • MDL number MFCD09056756
  • Cas Number : 133720-60-2
  • Molecular Weight : 197.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 7-bromo-1-benzofuran
Inchi Code 1S/C8H5BrO/c9-7-3-1-2-6-4-5-10-8(6)7/h1-5H
Inchi Key GPZPQJITKRSVQJ-UHFFFAOYSA-N

Description

Synonym 7-Bromo-1-benzofuran
Purity 95%
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/7-bromobenzobfuran

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/7-bromobenzobfuran
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart