APO455827816 Apollo Scientific Ltd

Methyl 1-phenylcyclobutane-1-carboxylate

  • MDL number MFCD11035753
  • Cas Number : 58469-03-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-(Methoxycarbonyl)-1-phenylcyclobutane
Storage temp. Ambient temperature
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=43436

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=43436
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart