APO455827819 Apollo Scientific Ltd

1-[2-(Diphenylmethoxy)ethyl]homopiperazine

  • MDL number MFCD00729027
  • Cas Number : 150557-09-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-[2-(Benzhydryloxy)ethyl]homopiperazine, 1-[2-(Diphenylmethoxy)ethyl]-1,4-diazepane
Storage temp. Ambient temperature
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Boiling point 183/0.5mm
Product Link http://www.apolloscientific.co.uk/display_item.php?id=12906

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=12906
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart