APO455827835 Apollo Scientific Ltd

4-Amino-2-chloro-5-nitropyridine

  • MDL number MFCD09033797
  • Cas Number : 2604-39-9
  • Molecular Weight : 173.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-chloro-5-nitro-4-pyridinamine
Inchi Code 1S/C5H4ClN3O2/c6-5-1-3(7)4(2-8-5)9(10)11/h1-2H,(H2,7,8)
Inchi Key YKWBEPUOVBMENG-UHFFFAOYSA-N

Description

Synonym 2-Chloro-5-nitropyridin-4-amine
Purity 98%
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-amino-2-chloro-5-nitropyridine

Safety Information

signalword Warning
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-amino-2-chloro-5-nitropyridine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart