APO455827836 Apollo Scientific Ltd

2,4-Dichloro-5-nitropyridine

  • MDL number MFCD07368834
  • Cas Number : 4487-56-3
  • Molecular Weight : 192.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2,4-dichloro-5-nitropyridine
Inchi Code 1S/C5H2Cl2N2O2/c6-3-1-5(7)8-2-4(3)9(10)11/h1-2H
Inchi Key RZVJQUMDJUUBBF-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/24-dichloro-5-nitropyridine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/24-dichloro-5-nitropyridine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart