APO455827838 Apollo Scientific Ltd

Methyl 4-hydroxypyrimidine-5-carboxylate

  • MDL number MFCD01632117
  • Cas Number : 4774-35-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 209-210
Synonym 4-Hydroxy-5-(methoxycarbonyl)pyrimidine, 4-Hydroxy-5-(methoxycarbonyl)-1,3-diazine
Purity 95
Storage temp. Ambient temperature
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=43034

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=43034
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart