APO455827847 Apollo Scientific Ltd

Methyl 2-bromofuran-3-carboxylate

  • MDL number MFCD09879944
  • Cas Number : 197846-06-3
  • Molecular Weight : 205.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 49-52
Iupac Name methyl 2-bromo-3-furoate
Inchi Code 1S/C6H5BrO3/c1-9-6(8)4-2-3-10-5(4)7/h2-3H,1H3
Inchi Key XXUUZVIMSVBWIQ-UHFFFAOYSA-N

Description

Synonym Methyl 2-bromo-3-furoate
Purity 97%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 49-52
Product Link https://store.apolloscientific.co.uk/product/methyl-2-bromofuran-3-carboxylate

Safety Information

signalword Warning
pcodes P261;P280;P312
MSDS https://store.apolloscientific.co.uk/product/methyl-2-bromofuran-3-carboxylate
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart