APO455827861 Apollo Scientific Ltd

1-(Methylsulphonyl)piperidin-4-one

  • MDL number MFCD08061935
  • Cas Number : 218780-53-1
  • Molecular Weight : 177.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-(Methylsulphonyl)-4-oxopiperidine
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/1-methylsulphonylpiperidin-4-one

Safety Information

signalword Warning
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/1-methylsulphonylpiperidin-4-one
pictograms GHS07
hstatements H302+H312+H332
Availability details for
Quantity
  • Availability
  • Add to Cart