APO455827869 Apollo Scientific Ltd

4-Methyl-3-{[4-(pyridin-3-yl)pyrimidin-2-yl]amino}benzoic acid

  • MDL number MFCD11521324
  • Cas Number : 641569-94-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-[(5-Carboxy-2-methylphenyl)amino]-4-(pyridin-3-yl)pyrimidine, 3-{2-[(5-Carboxy-2-methylphenyl)amino]pyrimidin-4-yl}pyridine
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/4-methyl-3-4-pyridin-3-ylpyrimidin-2-ylaminobenzoic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-methyl-3-4-pyridin-3-ylpyrimidin-2-ylaminobenzoic-acid
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart