APO455827878 Apollo Scientific Ltd

N-(2-Chloroethyl)-1H-pyrazole-1-carboxamide

  • MDL number MFCD11109511
  • Cas Number : 1209343-75-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-[(2-Chloroethyl)carbamoyl]-1H-pyrazole, 1-{[(2-Chloroethyl)amino]carbonyl}-1H-pyrazole
Storage temp. Ambient temperature
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=46015

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=46015
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart