APO455827881 Apollo Scientific Ltd

Naphth-1-ylglyoxal hydrate

  • MDL number MFCD08705867
  • Cas Number : 16208-20-1
  • Molecular Weight : 202.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Flash Point 196
Storage temp. Store at 2-8°C
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Boiling point 401/760mm
Product Link https://store.apolloscientific.co.uk/product/naphth-1-ylglyoxal-hydrate

Safety Information

signalword Warning
pcodes P261;P280;P301+P312
MSDS https://store.apolloscientific.co.uk/product/naphth-1-ylglyoxal-hydrate
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart