APO455827882 Apollo Scientific Ltd

(5-Bromothien-2-yl)glyoxal hydrate

  • MDL number MFCD08705872
  • Cas Number : 852619-28-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (5-Bromothien-2-yl)(oxo)acetaldehyde hydrate, 1-(5-Bromothien-2-yl)-2,2-dihydroxyethan-1-one
Purity 95
Storage temp. Store at 2-8
Physical Form Solid-Powder
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=29537

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=29537
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart