APO455827892 Apollo Scientific Ltd

Methyl (2S,4R)-4-aminopyrrolidine-2-carboxylate hydrochloride, N1-BOC protected

  • MDL number MFCD03787929
  • Cas Number : 334999-32-5
  • Molecular Weight : 280.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-tert-butyl 2-methyl (2S,4R)-4-amino-1,2-pyrrolidinedicarboxylate hydrochloride
Inchi Code 1S/C11H20N2O4.ClH/c1-11(2,3)17-10(15)13-6-7(12)5-8(13)9(14)16-4;/h7-8H,5-6,12H2,1-4H3;1H/t7-,8+;/m1./s1
Inchi Key KFYCQKLSGMAVQH-WLYNEOFISA-N

Description

Synonym 1-tert-Butyl 2-methyl (2S,4R)-4-aminopyrrolidine-1,2-dicarboxylate hydrochloride
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/methyl-2s4r-4-aminopyrrolidine-2-carboxylate-hydrochloride-n1-boc-protected

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-2s4r-4-aminopyrrolidine-2-carboxylate-hydrochloride-n1-boc-protected
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart