APO455827911 Apollo Scientific Ltd

Methyl 3,4-dihydro-2H-1,4-benzoxazine-7-carboxylate

  • MDL number MFCD11848180
  • Cas Number : 142166-01-6
  • Molecular Weight : 193.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 86-88
Iupac Name methyl 3,4-dihydro-2H-1,4-benzoxazine-7-carboxylate
Inchi Code 1S/C10H11NO3/c1-13-10(12)7-2-3-8-9(6-7)14-5-4-11-8/h2-3,6,11H,4-5H2,1H3
Inchi Key QHAIQLAFSPRIGM-UHFFFAOYSA-N

Description

Synonym 3,4-Dihydro-7-(methoxycarbonyl)-2H-1,4-benzoxazine, 3,4-Dihydro-7-(methoxycarbonyl)-2H-benzo[b][1,4]oxazine
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 86-88
Product Link https://store.apolloscientific.co.uk/product/methyl-34-dihydro-2h-14-benzoxazine-7-carboxylate

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-34-dihydro-2h-14-benzoxazine-7-carboxylate
pictograms GHS07
hstatements H302+H312+H332
Availability details for
Quantity
  • Availability
  • Add to Cart