APO455827918 Apollo Scientific Ltd

2-(Methylsulphonyl)aniline

  • MDL number MFCD08444390
  • Cas Number : 2987-49-7
  • Molecular Weight : 171.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 84-85
Synonym 2-Aminophenyl methyl sulphone
Purity 99
Physical Form Solid-Crystals
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-methylsulphonylaniline

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-methylsulphonylaniline
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart