APO455827921 Apollo Scientific Ltd

Benzo[b]furan-3-boronic acid, pinacol ester

  • MDL number MFCD11841067
  • Cas Number : 796851-30-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 68-70
Synonym 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1-benzofuran, 2-(1-Benzofuran-3-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=45677

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=45677
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart