APO455827924 Apollo Scientific Ltd

4'-(Benzyloxy)acetophenone

  • MDL number MFCD00017247
  • Cas Number : 54696-05-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 91-92
Synonym 1-[4-(Benzyloxy)phenyl]ethan-1-one, [(4-Acetylphenoxy)methyl]benzene
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=13021

Safety Information

signalword Warning
pcodes P273;P391;P501
MSDS http://www.apolloscientific.co.uk/display_item.php?id=13021
pictograms GHS09
hstatements H400
Availability details for
Quantity
  • Availability
  • Add to Cart