APO455827926 Apollo Scientific Ltd

6-Bromochroman

  • MDL number MFCD10698725
  • Cas Number : 3875-78-3
  • Molecular Weight : 213.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 6-Bromo-3,4-dihydro-2H-chromene
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin GB
Boiling point 82-84/0.05mm
Product Link https://store.apolloscientific.co.uk/product/6-bromochroman

Safety Information

signalword Warning
pcodes P271;P280;P260
MSDS https://store.apolloscientific.co.uk/product/6-bromochroman
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart