APO455827927 Apollo Scientific Ltd

4-(4-Bromo-1,3-thiazol-2-yl)morpholine

  • MDL number MFCD09607696
  • Cas Number : 1017781-60-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 74-76
Synonym 4-Bromo-2-(morpholin-4-yl)-1,3-thiazole
Purity 97
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/4-4-bromo-13-thiazol-2-ylmorpholine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-4-bromo-13-thiazol-2-ylmorpholine
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart